Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysine, N6-(1-oxo-2-buten-1-yl) | 1338823-35-0 | MFCD22381072 | 99.4% | 214.26 g/mol | C10H18N2O3 | 500 MG
SDP

Catalog No. 5000210109
Encompass_Preferred

N6-(But-2-enoyl)-L-lysine is an N6-acylated lysine derivative provided for research use as an amino acid derivative reagent. It is a white to off-white solid of high purity used in biochemical and synthetic chemistry applications where an N6-protected lysine analogue is required.

  • High purity of 99.4%.
  • Molecular weight 214.26 g/mol.
  • Chemical formula C10H18N2O3.
  • Available in multiple sizes, including 500 mg.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Relevant documentation available: data sheet, certificate of analysis, SDS, HNMR, LCMS.

Catalog No. 50-002-10109 Supplier Medchemexpress LLC Supplier No. HYW048203500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-(But-2-enoyl)-L-lysine is an N6-acylated lysine derivative provided for research use as an amino acid derivative reagent. It is a white to off-white solid of high purity used in biochemical and synthetic chemistry applications where an N6-protected lysine analogue is required.

  • High purity of 99.4%.
  • Molecular weight 214.26 g/mol.
  • Chemical formula C10H18N2O3.
  • Available in multiple sizes, including 500 mg.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Relevant documentation available: data sheet, certificate of analysis, SDS, HNMR, LCMS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.