Learn More
Medchemexpress LLC L-lysine, N6-[4-(4-iodophenyl)-1-oxobutyl]-, 1,1-dimethylethyl ester | 2703051-80-1 | 98.1% | 474.38 | C20H31IN2O3 | 10 MG
SDP
Catalog No. 5000288028
Encompass_Preferred
A lysine-based pharmacokinetic (PK) modifier supplied as a small-mass tert-butyl ester for use in the synthesis of PSMA-targeting ligands. It is designed to improve pharmacokinetic properties such as plasma residence time and to reduce off-target uptake, making it useful for targeted imaging and radionuclide therapy research.
  • Improves plasma residence time by enhancing albumin binding.
  • Reduces salivary gland uptake to help minimize off-target accumulation.
  • Suitable for incorporation into PSMA-targeting ligand synthesis workflows.
  • High purity appropriate for research use (98.1%).
  • Supplied as a tert-butyl ester for convenient synthetic handling.
Catalog No. 50-002-88028 Supplier Medchemexpress LLC Supplier No. HY15812210MG
A lysine-based pharmacokinetic (PK) modifier supplied as a small-mass tert-butyl ester for use in the synthesis of PSMA-targeting ligands. It is designed to improve pharmacokinetic properties such as plasma residence time and to reduce off-target uptake, making it useful for targeted imaging and radionuclide therapy research.
  • Improves plasma residence time by enhancing albumin binding.
  • Reduces salivary gland uptake to help minimize off-target accumulation.
  • Suitable for incorporation into PSMA-targeting ligand synthesis workflows.
  • High purity appropriate for research use (98.1%).
  • Supplied as a tert-butyl ester for convenient synthetic handling.