Learn More
Medchemexpress LLC L-lysine, N6-[(9H-fluoren-9-yl)methoxycarbonyl]-N6-carbamimidoyl-, (S)- | 776277-76-0 | 99.3% | 410.47 | C22H26N4O4 | 100 G
SDP
Catalog No. 5000208752
Encompass_Preferred
N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-carbamimidoyl-L-lysine is an Fmoc-protected L-lysine derivative bearing a carbamimidoyl (guanidino) side chain, supplied as a solid high-purity research reagent used as a building block in peptide synthesis and biochemical studies.
  • Provided as a solid powder suitable for peptide synthesis and storage.
  • High purity, suitable for analytical and synthetic applications.
  • Contains a guanidino-like side chain for arginine-mimetic properties.
  • Stable under recommended cold storage conditions for long-term use.
Catalog No. 50-002-08752 Supplier Medchemexpress LLC Supplier No. HYW089229100G
N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-carbamimidoyl-L-lysine is an Fmoc-protected L-lysine derivative bearing a carbamimidoyl (guanidino) side chain, supplied as a solid high-purity research reagent used as a building block in peptide synthesis and biochemical studies.
  • Provided as a solid powder suitable for peptide synthesis and storage.
  • High purity, suitable for analytical and synthetic applications.
  • Contains a guanidino-like side chain for arginine-mimetic properties.
  • Stable under recommended cold storage conditions for long-term use.