Learn More
Bioworld L-LYSINE SEPAROPOREAGAROS
Supplier:  Bioworld 201810703
Encompass
L-LYSINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: L-Lysine; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B (Agarose 4% Beads); pH stability: 4-9; Flow specifications: 70-140 cm/h; ...
Catalog No. 50-197-848