Learn More
Medchemexpress LLC L-Lysine (standard) | 56-87-1 | 146.19 | 250 MG
SDP
Catalog No. 5000389935
Encompass_Preferred
L-Lysine (Standard) is the analytical standard of L-Lysine, intended for research and analytical applications. It is an essential amino acid for humans with oral activity. It can inhibit HSV infections, increase calcium absorption, reduce diabetes-related diseases, improve gut health, and alleviate pancreatic inflammation.
  • Inhibits HSV infections
  • Increases calcium absorption
  • Reduces diabetes-related diseases
  • Improves gut health
  • Alleviates pancreatic inflammation
  • Used in research on metabolism, infection, and inflammation
  • Analytical standard for HPLC, GC, and MS experiments
Catalog No. 50-003-89935 Supplier Medchemexpress LLC Supplier No. 250MGHYN0469R
L-Lysine (Standard) is the analytical standard of L-Lysine, intended for research and analytical applications. It is an essential amino acid for humans with oral activity. It can inhibit HSV infections, increase calcium absorption, reduce diabetes-related diseases, improve gut health, and alleviate pancreatic inflammation.
  • Inhibits HSV infections
  • Increases calcium absorption
  • Reduces diabetes-related diseases
  • Improves gut health
  • Alleviates pancreatic inflammation
  • Used in research on metabolism, infection, and inflammation
  • Analytical standard for HPLC, GC, and MS experiments