Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-α-lysophosphatidylinositol sodium salt | 796963-91-2 | MFCD22416797 | >99.0% | 594.605 g/mol | C25H48NaO12P | 1 MG
SDP

Supplier:  Medchemexpress LLC HYW0200351MG

Encompass_Preferred

L-α-lysophosphatidylinositol sodium salt (CAS 796963-91-2) is a soy-derived lysophospholipid research reagent and GPR55 agonist used in biochemical and cell-based signaling studies. It is supplied as a white, hygroscopic powder, typically packaged under inert gas and stored frozen to preserve activity.

  • Purity >99.0% by HPLC.
  • Empirical formula C25H48NaO12P.
  • White, hygroscopic powder packaged under inert gas.
  • Source: soy; contains mixed fatty-acyl species.
  • Recommended storage -20°C; protect from light and moisture.
  • Used as a GPR55 agonist in receptor and cell signaling assays.
  • Supplied in small quantities suitable for research use.

Catalog No. 50-003-00912


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.