Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysyl-L-cysteine trifluoroacetate | 97.8% | 477.38 g/mol | C13H21F6N3O7S | 5 MG
SDP

Catalog No. 5000264669
Encompass_Preferred

L-lysyl-L-cysteine trifluoroacetate is a dipeptide formed from lysine and cysteine, provided as a solid TFA salt for research use. The material is characterized by a reported purity, defined molecular formula and molecular weight, and specified storage recommendations for powder and solutions.

  • Dipeptide composed of lysine and cysteine.
  • Supplied as trifluoroacetate (TFA) salt in solid powder form.
  • Reported purity 97.8%.
  • Molecular formula C13H21F6N3O7S and molecular weight 477.38 g/mol.
  • Available in small research quantities: 5 mg, 10 mg, 25 mg.
  • Store sealed, away from moisture; powder: -80°C for 2 years, -20°C for 1 year; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-64669 Supplier Medchemexpress LLC Supplier No. HY114788A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-lysyl-L-cysteine trifluoroacetate is a dipeptide formed from lysine and cysteine, provided as a solid TFA salt for research use. The material is characterized by a reported purity, defined molecular formula and molecular weight, and specified storage recommendations for powder and solutions.

  • Dipeptide composed of lysine and cysteine.
  • Supplied as trifluoroacetate (TFA) salt in solid powder form.
  • Reported purity 97.8%.
  • Molecular formula C13H21F6N3O7S and molecular weight 477.38 g/mol.
  • Available in small research quantities: 5 mg, 10 mg, 25 mg.
  • Store sealed, away from moisture; powder: -80°C for 2 years, -20°C for 1 year; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.