Learn More
TARGETMOL CHEMICALS INC L-Lysyl-L-lysine dihydrochlori
SDP
Catalog No. 502876107
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. L-Lysyl-L-lysine dihydrochloride (Lysyllysine dihydrochloride) is an enzyme cleavable linker. L-Lysyl-L-lysine dihydrochloride can be used as facile linker for delivering multiple biologically active peptides.
Catalog No. 50-287-6107 Supplier TARGETMOL CHEMICALS INC Supplier No. T61156100MG
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. L-Lysyl-L-lysine dihydrochloride (Lysyllysine dihydrochloride) is an enzyme cleavable linker. L-Lysyl-L-lysine dihydrochloride can be used as facile linker for delivering multiple biologically active peptides.