Learn More
Medchemexpress LLC L-lysyl-L-phenylalanine | 6235-35-4 | 96.8% | C15H23N3O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13551410MG
Encompass_Preferred
Lys-Phe (L-lysyl-L-phenylalanine) is a synthetic dipeptide supplied for research use. It has been reported to inhibit cell sickling and the gelation of sickle-cell hemoglobin solutions. Supplied as a powder or in solution formats, the compound is provided in small research-scale quantities with recommended low-temperature storage to preserve stability.
  • Inhibits cell sickling and hemoglobin gelation.
  • High purity (96.8%).
  • Available in multiple small package sizes suitable for laboratory studies.
  • Recommended storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.
Catalog No. 50-002-59965