Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Malate Dehydrogenase L-M
Catalog No. 502901551
Encompass
L-malateNAD oxidoreductaseThe L-malate dehydrogenase enzyme is a nuclear gene product that is synthesized with a 24-residue amino-terminal signal peptide. This peptide is proteolytically cleaved during the translocation of the enzyme to the mitochondrial matrix.
Catalog No. 50-290-1551 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127914001
L-malateNAD oxidoreductaseThe L-malate dehydrogenase enzyme is a nuclear gene product that is synthesized with a 24-residue amino-terminal signal peptide. This peptide is proteolytically cleaved during the translocation of the enzyme to the mitochondrial matrix.