Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(-)-Malic Acid, Spectrum™ Chemical
SDP

Catalog No. 18621082 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-082 25 g
18-621-083 100 g
18-621-084 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-621-082 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M205025GM
Add to Cart
Add to Cart

Synonym: (S)-2-Hydroxysuccinic Acid, (2S)-2-Hydroxybutanedioic Acid, L-Malic Acid

L-(-)-Malic Acid is a dicarboxylic acid and organic compound made by all living organisms. Malic acid is responsible for the sour taste of most fruits and is utilized as a food additive. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 97-67-6

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.