Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Mannitol | 643-01-6 | 99.80% | 182.17 | 50 MG
SDP

Catalog No. 5000350100
Encompass_Preferred

L-mannitol is a compound that can be used for compound sweetener synthesis. It is for research use only.

  • Molecular Formula: C₆H₁₄O₆
  • Molecular Weight: 182.17
  • Purity: 99.80%
  • CAS No.: 643-01-6
  • Appearance: White to off-white solid
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-50100 Supplier Medchemexpress LLC Supplier No. HY13931250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-mannitol is a compound that can be used for compound sweetener synthesis. It is for research use only.

  • Molecular Formula: C₆H₁₄O₆
  • Molecular Weight: 182.17
  • Purity: 99.80%
  • CAS No.: 643-01-6
  • Appearance: White to off-white solid
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.