Learn More
Medchemexpress LLC L-mannose | 10030-80-5 | MFCD00063884 | 98.0% | 180.16 g/mol | C6H12O6 | 100 MG
SDP
Catalog No. 5000303325
Encompass_Preferred
L-mannose is a naturally occurring monosaccharide supplied as a solid reagent for laboratory research. It is used as a biochemical substrate and in enzyme assays, notably as a substrate for mannose-6-phosphate isomerase, and is suitable for metabolic and analytical studies. The compound has a molecular weight of 180.16 g/mol and is commonly provided at about 98% purity.
  • Naturally occurring monosaccharide reagent
  • High purity (≈98.0%) suitable for research use
  • Molecular weight 180.16 g/mol; formula C6H12O6
  • Supplied as a solid for ease of handling
  • Useful as a substrate for mannose-6-phosphate isomerase and enzyme assays
  • Suitable for biochemical and metabolic studies
Catalog No. 50-003-03325 Supplier Medchemexpress LLC Supplier No. HYW016508100MG
L-mannose is a naturally occurring monosaccharide supplied as a solid reagent for laboratory research. It is used as a biochemical substrate and in enzyme assays, notably as a substrate for mannose-6-phosphate isomerase, and is suitable for metabolic and analytical studies. The compound has a molecular weight of 180.16 g/mol and is commonly provided at about 98% purity.
  • Naturally occurring monosaccharide reagent
  • High purity (≈98.0%) suitable for research use
  • Molecular weight 180.16 g/mol; formula C6H12O6
  • Supplied as a solid for ease of handling
  • Useful as a substrate for mannose-6-phosphate isomerase and enzyme assays
  • Suitable for biochemical and metabolic studies