Learn More
Medchemexpress LLC L-Mannose | 10030-80-5 | MFCD00136021 | 98.0% | 180.16 g/mol | C6H12O6 | 25 MG
SDP
Catalog No. 5000302896
Encompass_Preferred
L-Mannose is a naturally occurring monosaccharide (C6H12O6, MW 180.16 g/mol) used as a substrate for mannose-6-phosphate isomerase and as a reagent in biochemical and enzymatic assays. Supplied as a solid for research use, this compound is accompanied by batch-specific documentation to support quality and reproducibility.
  • Monosaccharide substrate for mannose-6-phosphate isomerase assays.
  • High purity, typically 98.0% by NMR.
  • Available in small research quantities for assay development.
  • Solid form, suitable for standard laboratory storage.
  • Certificate of analysis available for each batch.
Catalog No. 50-003-02896 Supplier Medchemexpress LLC Supplier No. 25MGHYW016508
L-Mannose is a naturally occurring monosaccharide (C6H12O6, MW 180.16 g/mol) used as a substrate for mannose-6-phosphate isomerase and as a reagent in biochemical and enzymatic assays. Supplied as a solid for research use, this compound is accompanied by batch-specific documentation to support quality and reproducibility.
  • Monosaccharide substrate for mannose-6-phosphate isomerase assays.
  • High purity, typically 98.0% by NMR.
  • Available in small research quantities for assay development.
  • Solid form, suitable for standard laboratory storage.
  • Certificate of analysis available for each batch.