Learn More
Medchemexpress LLC L-mannose | 10030-80-5 | MFCD00136021 | 98.0% | 180.16 g·mol⁻¹ | C6H12O6 | 50 MG
SDP
Catalog No. 5000303397
Encompass_Preferred
L-Mannose is a naturally occurring monosaccharide (CAS 10030-80-5) supplied as a solid reagent for biochemical research. It has formula C6H12O6, molecular weight 180.16 g·mol⁻¹, and is commonly used as a substrate in enzymatic assays and stereochemical carbohydrate studies.
  • Monosaccharide reagent suitable for enzymatic assays.
  • Purity 98.0% for reproducible results.
  • Solid form, easy to weigh and store.
  • Useful as a substrate for mannose-6-phosphate isomerase studies.
  • Available in small pack sizes for research-scale use.
Catalog No. 50-003-03397 Supplier Medchemexpress LLC Supplier No. HYW01650850MG
L-Mannose is a naturally occurring monosaccharide (CAS 10030-80-5) supplied as a solid reagent for biochemical research. It has formula C6H12O6, molecular weight 180.16 g·mol⁻¹, and is commonly used as a substrate in enzymatic assays and stereochemical carbohydrate studies.
  • Monosaccharide reagent suitable for enzymatic assays.
  • Purity 98.0% for reproducible results.
  • Solid form, easy to weigh and store.
  • Useful as a substrate for mannose-6-phosphate isomerase studies.
  • Available in small pack sizes for research-scale use.