Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-menthyl acetate | 2623-23-6 | 5 G
SDP

Catalog No. 5000419989
Encompass_Preferred

L-Menthyl acetate is a derivative of L-menthol, effective in enhancing 5-aminolevulinic acid (ALA) skin permeation.

  • Purity of 99.86%
  • Liquid appearance
  • Colorless to off-white color
  • Molecular weight of 198.30
  • Chemical formula C12H22O2
  • Soluble in DMSO at 100 mg/mL (504.29 mM)

Catalog No. 50-004-19989 Supplier Medchemexpress LLC Supplier No. HYN71325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Menthyl acetate is a derivative of L-menthol, effective in enhancing 5-aminolevulinic acid (ALA) skin permeation.

  • Purity of 99.86%
  • Liquid appearance
  • Colorless to off-white color
  • Molecular weight of 198.30
  • Chemical formula C12H22O2
  • Soluble in DMSO at 100 mg/mL (504.29 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.