Learn More
Medchemexpress LLC HY-N0326 1g Medchemexpress, L-Methionin CAS:63-68-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN03261G
Encompass_Preferred
Medchemexpress, HY-N0326 1g L-Methionin CAS:63-68-3 L-Methionine is the L-isomer of Methionine, an essential amino acid for human development. Methionine acts as a hepatoprotectant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3522