Learn More
Cayman Chemical L-Methionine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 13010-53-2, 100 mg
SDP
Catalog No. 502912270
Encompass_Preferred
An internal standard for the quantification of L-methionine by GC- or LC-MS
Catalog No. 50-291-2270 Supplier Cayman Chemical Supplier No. 34826100 MG
An internal standard for the quantification of L-methionine by GC- or LC-MS