Learn More
Cayman Chemical L-MethIonIn-d3 1g
SDP
Catalog No. 502912736
Encompass_Preferred
An internal standard for the quantification of L-methionine by GC- or LC-MS
Catalog No. 50-291-2736 Supplier Cayman Chemical Supplier No. 348261 G
Add to Cart
Add to Cart
An internal standard for the quantification of L-methionine by GC- or LC-MS