Learn More
Cayman Chemical L-Methionine-(S,R)-Sulfoximine, Purity: ≥95%, CAS: 15985-39-4, 100 mg
SDP
Catalog No. 502937858
Encompass_Preferred
An inhibitor of glutamine synthetase; inhibits pea glutamine synthetase by 31-64% at 5-25 µM following preincubation with L-glutamine; inhibits glutamine synthetase in isolated chick embryo retinas
Catalog No. 50-293-7858 Supplier Cayman Chemical Supplier No. 27608100 MG
An inhibitor of glutamine synthetase; inhibits pea glutamine synthetase by 31-64% at 5-25 µM following preincubation with L-glutamine; inhibits glutamine synthetase in isolated chick embryo retinas