Learn More
Cayman Chemical L-Methionine-(S,R)-Sulfoximine, Purity: ≥95%, CAS: 15985-39-4, 1 g
SDP
Catalog No. 502941567
Encompass_Preferred
An inhibitor of glutamine synthetase; inhibits pea glutamine synthetase by 31-64% at 5-25 µM following preincubation with L-glutamine; inhibits glutamine synthetase in isolated chick embryo retinas
Catalog No. 50-294-1567 Supplier Cayman Chemical Supplier No. 276081 G
An inhibitor of glutamine synthetase; inhibits pea glutamine synthetase by 31-64% at 5-25 µM following preincubation with L-glutamine; inhibits glutamine synthetase in isolated chick embryo retinas