Learn More
Cayman Chemical L-MethIonIn SulfoxIde 1g
SDP
Catalog No. 502912371
Encompass_Preferred
A sulfoxide-modified methionine; increases weight gain in weanling mice in the diet at 6.3, 12.6, and 18.9 mmol/g; levels are decreased in patients with vitiligo
Catalog No. 50-291-2371 Supplier Cayman Chemical Supplier No. 362551 G
A sulfoxide-modified methionine; increases weight gain in weanling mice in the diet at 6.3, 12.6, and 18.9 mmol/g; levels are decreased in patients with vitiligo