Learn More
Medchemexpress LLC L-methioninamide hydrochloride | 16120-92-6 | MFCD00039084 | 99.2% | 184.69 g·mol⁻¹ | C5H13ClN2OS | 1 ML
SDP
Catalog No. 5000303719
Encompass_Preferred
L-methioninamide hydrochloride is the hydrochloride salt of L-methioninamide, a methionine analogue reported to inhibit methionyl-tRNA synthetase. It is supplied for research use as a solid or as a 10 mM solution in DMSO; not for human or clinical use.
  • Acts as a methionyl-tRNA synthetase inhibitor for biochemical studies.
  • Available as powder and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research applications.
  • White to off-white solid with defined storage recommendations.
  • Useful in studies of methionine metabolism and protein synthesis.
Catalog No. 50-003-03719 Supplier Medchemexpress LLC Supplier No. HYW01625610MM/1ML
L-methioninamide hydrochloride is the hydrochloride salt of L-methioninamide, a methionine analogue reported to inhibit methionyl-tRNA synthetase. It is supplied for research use as a solid or as a 10 mM solution in DMSO; not for human or clinical use.
  • Acts as a methionyl-tRNA synthetase inhibitor for biochemical studies.
  • Available as powder and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research applications.
  • White to off-white solid with defined storage recommendations.
  • Useful in studies of methionine metabolism and protein synthesis.