Learn More
Medchemexpress LLC L-methioninamide hydrochloride | 16120-92-6 | MFCD00039084 | 99.2% | 184.69 g/mol | C5H13ClN2OS | 10 G
SDP
Catalog No. 5000302310
Encompass_Preferred
L-methioninamide hydrochloride is the hydrochloride salt of L-methioninamide, a methionine analogue reported to inhibit methionyl-tRNA synthetase. It is supplied for research use and is available as solid material or as ready-to-use DMSO solutions for biochemical and biological studies.
  • Acts as a methionyl-tRNA synthetase inhibitor.
  • Available as solid and as 10 mM DMSO solution formats.
  • High purity suitable for analytical and biological research.
  • Molecular weight 184.69 g/mol; formula C5H13ClN2OS.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-02310 Supplier Medchemexpress LLC Supplier No. HYW01625610G
L-methioninamide hydrochloride is the hydrochloride salt of L-methioninamide, a methionine analogue reported to inhibit methionyl-tRNA synthetase. It is supplied for research use and is available as solid material or as ready-to-use DMSO solutions for biochemical and biological studies.
  • Acts as a methionyl-tRNA synthetase inhibitor.
  • Available as solid and as 10 mM DMSO solution formats.
  • High purity suitable for analytical and biological research.
  • Molecular weight 184.69 g/mol; formula C5H13ClN2OS.
  • For research use only; not for human or clinical use.