Learn More
Research Products International Corp L-Methionine, 100 Grams
SDP
Supplier:  Research Products International Corp M22060100.0
Encompass_Preferred
L-Methionine is a key participant in protein synthesis, serving as the initiator amino acid for the translation of mRNA into a polypeptide chain.
It is the starting point for the synthesis of proteins within cells and is involved in methylation reactions, where it serves as a donor of methyl groups. Methylation is a crucial process in the regulation of gene expression and other cellular functions. 
It has antioxidant properties and can contribute to the cellular defense against oxidative stress. Also, it helps neutralize free radicals and reduce oxidative damage to cells.
L-Methionine is a common component in laboratory research, particularly in studies related to biochemistry, molecular biology, and cell biology. Its significance as an essential amino acid makes it valuable for understanding cellular processes.
Catalog No. 50-213-449