Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-methionine-13C5,15N | 202468-47-1 | 99.2% | 155.17 g·mol⁻¹ | 13C5H11 15NO2S | 10 MG
SDP

Catalog No. 5000295809
Encompass_Preferred

L-methionine-13C5,15N is the stable isotope-labeled form of the essential amino acid L-methionine supplied as a powder for use as an internal standard, tracer, or reference in mass spectrometry and metabolic studies.

  • Contains five 13C atoms and one 15N atom.
  • Purity 99.2% by assay.
  • Molecular weight 155.17 g·mol⁻1; molecular formula 13C5H11 15NO2S.
  • CAS number 202468-47-1 for unambiguous identification.
  • Stored as powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in small-mass packages suitable for analytical use, including 10 mg.

Catalog No. 50-002-95809 Supplier Medchemexpress LLC Supplier No. HYN0326S610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-methionine-13C5,15N is the stable isotope-labeled form of the essential amino acid L-methionine supplied as a powder for use as an internal standard, tracer, or reference in mass spectrometry and metabolic studies.

  • Contains five 13C atoms and one 15N atom.
  • Purity 99.2% by assay.
  • Molecular weight 155.17 g·mol⁻1; molecular formula 13C5H11 15NO2S.
  • CAS number 202468-47-1 for unambiguous identification.
  • Stored as powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in small-mass packages suitable for analytical use, including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.