Learn More
Selleck Chemical LLC L-methionine
SDP
Catalog No. 5000004012
Encompass_Preferred
L-Methionine is an essential sulfur-containing free-form amino acid acts as an antioxidant promoter and contributes to the synthesis of S-adenosyl-L-methionine (SAMe)
Catalog No. 50-000-04012 Supplier Selleck Chemical LLC Supplier No. S5633100MG
L-Methionine is an essential sulfur-containing free-form amino acid acts as an antioxidant promoter and contributes to the synthesis of S-adenosyl-L-methionine (SAMe)