Learn More
Medchemexpress LLC L-Methionine | 63-68-3 | 99.8% | 149.21 | 1 ML
SDP
Catalog No. 5000427498
Encompass_Preferred
L-Methionine is an L-isomer of orally active Methionine, an essential amino acid. It functions as a strong liver antidote and liver protector. L-Methionine can inhibit cell proliferation, induce cell apoptosis, and possesses antitumor and antioxidant activity.
  • Inhibits cell proliferation
  • Induces cell apoptosis
  • Functions as a strong liver antidote and protector
  • Possesses antitumor activity
  • Possesses antioxidant activity
Catalog No. 50-004-27498 Supplier Medchemexpress LLC Supplier No. HYN032610MM/1ML
L-Methionine is an L-isomer of orally active Methionine, an essential amino acid. It functions as a strong liver antidote and liver protector. L-Methionine can inhibit cell proliferation, induce cell apoptosis, and possesses antitumor and antioxidant activity.
  • Inhibits cell proliferation
  • Induces cell apoptosis
  • Functions as a strong liver antidote and protector
  • Possesses antitumor activity
  • Possesses antioxidant activity