Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-methionine 7-amido-4-methylcoumarin (trifluoroacetate) | 94367-35-8 | 99.9% | 420.40 | C17H19F3N2O5S | 10 MG
SDP

Catalog No. 5000209331
Encompass_Preferred

L-methionine 7-amido-4-methylcoumarin (trifluoroacetate) is a biochemical assay reagent supplied as a white to off-white solid for research use. It is used as a fluorogenic substrate in enzymatic assays and analytical applications, and is provided in milligram pack sizes with specified storage recommendations.

  • White to off-white solid with 99.9% purity.
  • Molecular formula C17H19F3N2O5S and molecular weight 420.40.
  • Trifluoroacetate salt form.
  • Pack size 10 mg.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09331 Supplier Medchemexpress LLC Supplier No. HYW04028010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-methionine 7-amido-4-methylcoumarin (trifluoroacetate) is a biochemical assay reagent supplied as a white to off-white solid for research use. It is used as a fluorogenic substrate in enzymatic assays and analytical applications, and is provided in milligram pack sizes with specified storage recommendations.

  • White to off-white solid with 99.9% purity.
  • Molecular formula C17H19F3N2O5S and molecular weight 420.40.
  • Trifluoroacetate salt form.
  • Pack size 10 mg.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.