Learn More
Medchemexpress LLC L-Methionine-DL-sulfoximine | 15985-39-4 | 98.0% | 1 G
SDP
Catalog No. 5000411829
Encompass_Preferred
L-Methionine-DL-sulfoximine (MSX; MSO) is a highly specific and irreversible inhibitor of Glutamine synthetase (GS). It acts as a potent convulsant, primarily affecting astroglia metabolically and morphologically. This compound has been utilized to inhibit Gln-dependent ammonia-stimulated neuronal toxicity in vitro and significantly increases the release rate of fixed nitrogen in cyanobacteria. It is considered a promising candidate for research in biofertilizers and convulsive seizures (CS).
  • Highly specific and irreversible inhibitor of glutamine synthetase.
  • Potent convulsant affecting astroglia.
  • Inhibits Gln-dependent ammonia-stimulated neuronal toxicity in vitro.
  • Increases release rate of fixed nitrogen in cyanobacteria.
  • Potential for research in biofertilizers and convulsive seizures.
Catalog No. 50-004-11829 Supplier Medchemexpress LLC Supplier No. HYB16921G
L-Methionine-DL-sulfoximine (MSX; MSO) is a highly specific and irreversible inhibitor of Glutamine synthetase (GS). It acts as a potent convulsant, primarily affecting astroglia metabolically and morphologically. This compound has been utilized to inhibit Gln-dependent ammonia-stimulated neuronal toxicity in vitro and significantly increases the release rate of fixed nitrogen in cyanobacteria. It is considered a promising candidate for research in biofertilizers and convulsive seizures (CS).
  • Highly specific and irreversible inhibitor of glutamine synthetase.
  • Potent convulsant affecting astroglia.
  • Inhibits Gln-dependent ammonia-stimulated neuronal toxicity in vitro.
  • Increases release rate of fixed nitrogen in cyanobacteria.
  • Potential for research in biofertilizers and convulsive seizures.