Learn More
Medchemexpress LLC L-Methionine-DL-sulfoximine (MSX; MSO) | 15985-39-4 | MFCD00002621 | 98.0% | 1 ML
SDP
Catalog No. 5000413285
Encompass_Preferred
L-Methionine-DL-sulfoximine (MSX; MSO) is a highly specific and irreversible inhibitor of Glutamine synthetase (GS). It is also a potent convulsant that primarily affects astroglia metabolically and morphologically. This product is for research use only.
  • Inhibits Gln-dependent ammonia-stimulated neuronal toxicity in vitro
  • Potentiates Gln deficit-dependent depression
  • Significantly increases fixed nitrogen release in cyanobacteria
  • Promising candidate for research in biofertilizers
  • Promising candidate for research in convulsive seizures
Catalog No. 50-004-13285 Supplier Medchemexpress LLC Supplier No. HYB169210MM/1ML
L-Methionine-DL-sulfoximine (MSX; MSO) is a highly specific and irreversible inhibitor of Glutamine synthetase (GS). It is also a potent convulsant that primarily affects astroglia metabolically and morphologically. This product is for research use only.
  • Inhibits Gln-dependent ammonia-stimulated neuronal toxicity in vitro
  • Potentiates Gln deficit-dependent depression
  • Significantly increases fixed nitrogen release in cyanobacteria
  • Promising candidate for research in biofertilizers
  • Promising candidate for research in convulsive seizures