Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Methionine-(methyl-13C) 99 atom % 13C | 49705-26-2 | MFCD00002623 | 1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2991461G
Encompass
L-Methionine-(methyl-13C) 99 atom % 13C | Purity: 99% (CP) | Mol Wt: 150.2 | 49705-26-2 | MFCD00002623 | 1G
Catalog No. 50-185-6022