Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine Methyl Ester Hydrochloride, 99+%, MP biomedicals™

Catalog No. ICN10229401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10229401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10229401 Supplier MP Biomedicals, Inc Supplier No. 0210229401

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2491-18-1
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00012491,MFCD00067540
InChI Key MEVUPUNLVKELNV-UHFFFAOYNA-N
Synonym h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
PubChem CID 11435579
IUPAC Name hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
SMILES [H+].[Cl-].COC(=O)C(N)CCSC

Specifications

Color White
Assay Percent Range 99+%
Quantity 1 g
Formula Weight 199.7
Physical Form Powder
Chemical Name or Material L-Methionine Methyl Ester Hydrochloride, For TLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.