Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ L-Methionine (Methyl-13C)

Catalog No. A39248
Encompass_Preferred
Change view
Click to view available options
Quantity:
225 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39248 225 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39248 Supplier Gibco™ Supplier No. A39248
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Methionine is a common sulfur-containing amino acid that is commonly used for the isotopic labeling of methionine protein residues for NMR spectroscopy studies.

Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.

EC Number: 200-562-9 (unlabeled)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-2-Amino-4-(Methylmercapto)butyric acid / L-2-Amino-4-(methylthio)cutanoic acid
Molecular Weight (g/mol) 150.2
Quantity 225 mg
Percent Purity >98%
Physical Form Powder
Shipping Condition Wet Ice
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.