Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine, MP Biomedicals™

Catalog No. MP021022915 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021022915 500 g
MP021022913 25 g
MP021022911 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP021022915 Supplier MP Biomedicals, Inc Supplier No. 02102291.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Methionine, MP Biomedicals is one of the common sulfur-containing amino acids. It is a methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol and choline.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 284°C (decomposition)(literature)
pH 5.6 to 6.1 (1% aq. soln.)
Quantity 500 g
Solubility Information Soluble in warm dil alcohol; insoluble in abs. alcohol, ether, petroleum ether, benzene, acetone, HCl.
Optical Rotation 22.4–24.7° (c=20, 6N HCl) (Lit.)
Formula Weight 149.21
Percent Purity ≥99%
Physical Form Crystalline Powder
Recommended Storage Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.