Learn More
Medchemexpress LLC L-methionine N-[(5-(imidazol-5-ylmethyl)amino)methyl-2'-methylbiphenyl-2-yl]carbonyl methyl ester | 344900-92-1 | 99.8% | 466.60 | C25H30N4O3S | 25MG
SDP
Catalog No. 5000239529
Encompass_Preferred
FTI-2153 is a potent, highly selective farnesyltransferase (FTase) inhibitor used in research to study Ras processing and mitotic spindle formation. It demonstrates low-nanomolar FTase inhibition and is applied in cell biology and oncology research to probe Ras-dependent signaling and mitotic effects.
  • Potent, selective inhibition of farnesyltransferase (IC50 ≈ 1.4 nM).
  • Inhibits H-Ras processing and affects mitotic spindle formation in cell assays.
  • Suitable for cell-based and biochemical studies of Ras signaling.
  • High purity appropriate for research applications (HPLC purity: 99.77%).
  • Supplied as a solid, convenient for storage and formulation.
  • Available in 25 MG quantities for laboratory use.
Catalog No. 50-002-39529 Supplier Medchemexpress LLC Supplier No. HY12324225MG
FTI-2153 is a potent, highly selective farnesyltransferase (FTase) inhibitor used in research to study Ras processing and mitotic spindle formation. It demonstrates low-nanomolar FTase inhibition and is applied in cell biology and oncology research to probe Ras-dependent signaling and mitotic effects.
  • Potent, selective inhibition of farnesyltransferase (IC50 ≈ 1.4 nM).
  • Inhibits H-Ras processing and affects mitotic spindle formation in cell assays.
  • Suitable for cell-based and biochemical studies of Ras signaling.
  • High purity appropriate for research applications (HPLC purity: 99.77%).
  • Supplied as a solid, convenient for storage and formulation.
  • Available in 25 MG quantities for laboratory use.