Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-methionine N-(5-(imidazol-5-ylmethylamino)methyl-2'-methylbiphenyl) methyl ester | 344900-92-1 | 99.8% | 466.60 g/mol | C25H30N4O3S | 10MG
SDP

Catalog No. 5000237024
Encompass_Preferred

FTI-2153 is a potent, highly selective farnesyltransferase inhibitor used as a research reagent to study protein prenylation, Ras processing, and cellular responses to FTase inhibition. The compound is provided with solubility and storage guidance to support preparation of DMSO stocks and in vitro or in vivo formulation workflows.

  • Potent FTase inhibition, IC50 1.4 nM.
  • Selective inhibition of H-Ras processing in cellular assays.
  • Available in small-mass vials and ready-to-use DMSO stock solutions.
  • Molecular weight 466.60 g/mol and formula C25H30N4O3S.
  • Storage recommended at -20°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO (up to ~90 mg/mL); datasheet provides stock preparation volumes and in vivo formulation options.

Catalog No. 50-002-37024 Supplier Medchemexpress LLC Supplier No. HY12324210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FTI-2153 is a potent, highly selective farnesyltransferase inhibitor used as a research reagent to study protein prenylation, Ras processing, and cellular responses to FTase inhibition. The compound is provided with solubility and storage guidance to support preparation of DMSO stocks and in vitro or in vivo formulation workflows.

  • Potent FTase inhibition, IC50 1.4 nM.
  • Selective inhibition of H-Ras processing in cellular assays.
  • Available in small-mass vials and ready-to-use DMSO stock solutions.
  • Molecular weight 466.60 g/mol and formula C25H30N4O3S.
  • Storage recommended at -20°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO (up to ~90 mg/mL); datasheet provides stock preparation volumes and in vivo formulation options.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.