Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-methionine p-nitroanilide | 6042-04-2 | MFCD00063424 | >98.0% | 269.32 g/mol | C11H15N3O3S | 5 G
SDP

Catalog No. 5000282987
Encompass_Preferred

L-methionine p-nitroanilide is an L-methionine derivative used as an amino-acid substrate in biochemical and enzymatic assays. It is provided as a light yellow solid intended for research use and requires low-temperature, light-protected storage to maintain stability.

  • Used as a substrate in enzyme assays and analytical applications.
  • Light yellow to yellow solid physical form.
  • Molecular weight 269.32 g/mol; chemical formula C11H15N3O3S.
  • CAS number 6042-04-2 for unambiguous identification.
  • Purity reported as ≥98% (TLC) by third-party suppliers.
  • Store at -20°C and protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-82987 Supplier Medchemexpress LLC Supplier No. HYW1419625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-methionine p-nitroanilide is an L-methionine derivative used as an amino-acid substrate in biochemical and enzymatic assays. It is provided as a light yellow solid intended for research use and requires low-temperature, light-protected storage to maintain stability.

  • Used as a substrate in enzyme assays and analytical applications.
  • Light yellow to yellow solid physical form.
  • Molecular weight 269.32 g/mol; chemical formula C11H15N3O3S.
  • CAS number 6042-04-2 for unambiguous identification.
  • Purity reported as ≥98% (TLC) by third-party suppliers.
  • Store at -20°C and protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.