Learn More
Apexbio Technology LLC L-Methionine sulfoxide 3226-65-1 500mg
SDP
Catalog No. 5000088473
Encompass_Preferred
L-Methionine sulfoxide (CAS 3226-65-1) is a small molecule derived from the oxidative metabolism of methionine It has been shown to promote polarization of macrophages toward the M1 phenotype implicating its influence on immune cell function Mechanistically L-Methionine sulfoxide modulates oxidative stress responses and purinergic signaling pathways This compound is utilized in experimental settings to investigate the regulation of redox homeostasis and immunity as well as to explore its impact on disease-relevant signaling networks and pathogenic processes
Catalog No. 50-000-88473 Supplier Apexbio Technology LLC Supplier No. C6651500
L-Methionine sulfoxide (CAS 3226-65-1) is a small molecule derived from the oxidative metabolism of methionine It has been shown to promote polarization of macrophages toward the M1 phenotype implicating its influence on immune cell function Mechanistically L-Methionine sulfoxide modulates oxidative stress responses and purinergic signaling pathways This compound is utilized in experimental settings to investigate the regulation of redox homeostasis and immunity as well as to explore its impact on disease-relevant signaling networks and pathogenic processes