Learn More
Medchemexpress LLC L-Methionine sulfoxide | 3226-65-1 | 98.77% | 165.21 | 50 G
SDP
Catalog No. 5000451618
Encompass_Preferred
L-Methionine sulfoxide is an orally active oxidation product of Methionine. It can be partially converted into Methionine within the body and participates in the synthesis of glutathione in the liver. It provides sulfur activity to the body and can participate in the synthesis of proteins and sulfur-containing compounds.
  • Can be partially converted into Methionine in the body.
  • Participates in the synthesis of glutathione in the liver.
  • Provides sulfur activity to the body.
  • Involved in the synthesis of proteins and sulfur-containing compounds.
  • Partially restores liver glutathione levels in male Wistar albino rats.
  • Approximately 60% efficient in utilizing methionine for growth in weanling rats.
Catalog No. 50-004-51618 Supplier Medchemexpress LLC Supplier No. HYW01010450G
L-Methionine sulfoxide is an orally active oxidation product of Methionine. It can be partially converted into Methionine within the body and participates in the synthesis of glutathione in the liver. It provides sulfur activity to the body and can participate in the synthesis of proteins and sulfur-containing compounds.
  • Can be partially converted into Methionine in the body.
  • Participates in the synthesis of glutathione in the liver.
  • Provides sulfur activity to the body.
  • Involved in the synthesis of proteins and sulfur-containing compounds.
  • Partially restores liver glutathione levels in male Wistar albino rats.
  • Approximately 60% efficient in utilizing methionine for growth in weanling rats.