Learn More
Medchemexpress LLC L-Methionine sulfoxide | 3226-65-1 | 98.8% | 165.21 | 100 G
SDP
Catalog No. 5000366428
Encompass_Preferred
L-Methionine sulfoxide is an orally active oxidation product of Methionine. It can be partially converted into Methionine within the body and contributes to the synthesis of glutathione in the liver. This product also supplies sulfur activity to the body, participating in the synthesis of proteins and various sulfur-containing compounds.
  • Orally active oxidation product of methionine.
  • Can be partially converted to methionine in the body.
  • Participates in the synthesis of glutathione in the liver.
  • Provides sulfur activity to the body.
  • Involved in the synthesis of proteins and sulfur-containing compounds.
Catalog No. 50-003-66428 Supplier Medchemexpress LLC Supplier No. HYW010104100G
L-Methionine sulfoxide is an orally active oxidation product of Methionine. It can be partially converted into Methionine within the body and contributes to the synthesis of glutathione in the liver. This product also supplies sulfur activity to the body, participating in the synthesis of proteins and various sulfur-containing compounds.
  • Orally active oxidation product of methionine.
  • Can be partially converted to methionine in the body.
  • Participates in the synthesis of glutathione in the liver.
  • Provides sulfur activity to the body.
  • Involved in the synthesis of proteins and sulfur-containing compounds.