Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Methionine sulfoxide | 3226-65-1 | 98.8% | 165.21 | 5 G
SDP

Catalog No. 5000453154
Encompass_Preferred

L-Methionine sulfoxide is an orally active oxidation product of Methionine. It can be partially converted into Methionine in the body and participates in the synthesis of glutathione in the liver. It provides the body with sulfur activity and can participate in the synthesis of proteins and sulfur-containing compounds.

  • Orally active oxidation product of Methionine.
  • Partially convertible to Methionine in the body.
  • Participates in glutathione synthesis in the liver.
  • Provides sulfur activity for protein and sulfur-containing compound synthesis.
  • For research use only.
  • Available in various solid quantities and as a solution.
  • Appearance: Solid, white to off-white.
  • Initial source: Endogenous metabolite.
  • Storage: 4°C, sealed storage, away from moisture and light; in solvent: -80°C, 6 months; -20°C, 1 month.

Catalog No. 50-004-53154 Supplier Medchemexpress LLC Supplier No. HYW0101045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Methionine sulfoxide is an orally active oxidation product of Methionine. It can be partially converted into Methionine in the body and participates in the synthesis of glutathione in the liver. It provides the body with sulfur activity and can participate in the synthesis of proteins and sulfur-containing compounds.

  • Orally active oxidation product of Methionine.
  • Partially convertible to Methionine in the body.
  • Participates in glutathione synthesis in the liver.
  • Provides sulfur activity for protein and sulfur-containing compound synthesis.
  • For research use only.
  • Available in various solid quantities and as a solution.
  • Appearance: Solid, white to off-white.
  • Initial source: Endogenous metabolite.
  • Storage: 4°C, sealed storage, away from moisture and light; in solvent: -80°C, 6 months; -20°C, 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.