Learn More
Apexbio Technology LLC L-(-)-α-Methyldopa (hydrate) 41372-08-1 1g
SDP
Catalog No. 5000090720
Encompass_Preferred
L-(-)- -Methyldopa (hydrate) (CAS 41372-08-1) is a small molecule that functions as an agonist selective for 2-adrenergic receptors Through its sympatholytic properties it diminishes central adrenergic outflow resulting in decreased blood pressure This compound is widely utilized in biomedical research to investigate mechanisms of adrenergic signaling and to model antihypertensive pharmacological effects Its ability to modulate neurotransmitter pathways makes it a useful tool for studies of cardiovascular regulation and central nervous system pharmacodynamics
Catalog No. 50-000-90720 Supplier Apexbio Technology LLC Supplier No. B10641000
L-(-)- -Methyldopa (hydrate) (CAS 41372-08-1) is a small molecule that functions as an agonist selective for 2-adrenergic receptors Through its sympatholytic properties it diminishes central adrenergic outflow resulting in decreased blood pressure This compound is widely utilized in biomedical research to investigate mechanisms of adrenergic signaling and to model antihypertensive pharmacological effects Its ability to modulate neurotransmitter pathways makes it a useful tool for studies of cardiovascular regulation and central nervous system pharmacodynamics