Learn More
Selleck Chemical LLC L-Mimosine 25mg 500-44-7 Leucenol
SDP
Catalog No. 501948071
Encompass_Preferred
L-Mimosine (Leucenol) is a non-protein amino acid, and acts as an iron chelator. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8071 Supplier Selleck Chemical LLC Supplier No. S744625MG
L-Mimosine (Leucenol) is a non-protein amino acid, and acts as an iron chelator. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.