Learn More
Cayman Chemical L-MoseshydrochlorIde 10mg
SDP
Catalog No. 502980912
Encompass_Preferred
A PCAF bromodomain inhibitor (Kd = 126 nM); selective for PCAF over GCN5 (Kd = 600 nM); >4,500-fold selective for PCAF over BRD4; displaces the PCAF bromodomain from histone H3.3 in a nanoBRET target engagement assay at 5 μM
Catalog No. 50-298-0912 Supplier Cayman Chemical Supplier No. 2128810 MG
A PCAF bromodomain inhibitor (Kd = 126 nM); selective for PCAF over GCN5 (Kd = 600 nM); >4,500-fold selective for PCAF over BRD4; displaces the PCAF bromodomain from histone H3.3 in a nanoBRET target engagement assay at 5 μM