Learn More
Selleck Chemical LLC L-NAME HCl 100mg 51298-62-5 NG-Nitroarginine methyl ester, N-Nitro-L-arginine methylester
SDP
Supplier:  Selleck Chemical LLC S2877100MG
Encompass_Preferred
L-NAME HCl (NG-Nitroarginine methyl ester, N-Nitro-L-arginine methylester) is a nonselective inhibitor of nitric oxide synthetases (NOS) for nNOS (bovine), eNOS (human), and iNOS (murine), with Ki of 15 nM, 39 nM and 4.4 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3126