Learn More
Medchemexpress LLC HY-18729A 100mg Medchemexpress, L-NAME (hydrochloride) CAS:51298-62-5 Purity:>98%
SDP
Catalog No. 502082079
Encompass_Preferred
Medchemexpress, HY-18729A 100mg L-NAME (hydrochloride) CAS:51298-62-5 L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. L-NAME is a precursor to NOS inhibitor L-NOARG which has an IC50 value of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2079 Supplier Medchemexpress LLC Supplier No. HY18729A100MG
Medchemexpress, HY-18729A 100mg L-NAME (hydrochloride) CAS:51298-62-5 L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. L-NAME is a precursor to NOS inhibitor L-NOARG which has an IC50 value of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.