Learn More
Medchemexpress LLC L-NAME (hydrochloride) | 51298-62-5 | 99.89% | 269.69 | 1 G
SDP
Catalog No. 5000387955
Encompass_Preferred
L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. L-NAME is a precursor to the NOS inhibitor L-NOARG, which has an IC50 value of 1.4 μM. This product can also be utilized to induce hypertension and preeclampsia models.
  • Inhibits NOS with an IC50 of 70 μM.
  • Functions as a precursor to the NOS inhibitor L-NOARG, which has an IC50 of 1.4 μM.
  • Applicable for inducing hypertension models.
  • Applicable for inducing preeclampsia models.
Catalog No. 50-003-87955 Supplier Medchemexpress LLC Supplier No. HY18729A1G
L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. L-NAME is a precursor to the NOS inhibitor L-NOARG, which has an IC50 value of 1.4 μM. This product can also be utilized to induce hypertension and preeclampsia models.
  • Inhibits NOS with an IC50 of 70 μM.
  • Functions as a precursor to the NOS inhibitor L-NOARG, which has an IC50 of 1.4 μM.
  • Applicable for inducing hypertension models.
  • Applicable for inducing preeclampsia models.