Learn More
Medchemexpress LLC L-NAME (hydrochloride) | 51298-62-5 | 99.9% | 269.69 | 25 G
SDP
Catalog No. 5000384818
Encompass_Preferred
L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. It is a precursor to the NOS inhibitor L-NOARG, which has an IC50 value of 1.4 μM. This product is intended for research use only and can be used to induce hypertension and preeclampsia models.
  • Inhibits NOS with an IC50 of 70 μM.
  • Functions as a precursor to the NOS inhibitor L-NOARG.
  • Suitable for inducing hypertension models.
  • Suitable for inducing preeclampsia models.
  • Exhibits a purity of 99.89%.
Catalog No. 50-003-84818 Supplier Medchemexpress LLC Supplier No. 25GHY18729A
L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. It is a precursor to the NOS inhibitor L-NOARG, which has an IC50 value of 1.4 μM. This product is intended for research use only and can be used to induce hypertension and preeclampsia models.
  • Inhibits NOS with an IC50 of 70 μM.
  • Functions as a precursor to the NOS inhibitor L-NOARG.
  • Suitable for inducing hypertension models.
  • Suitable for inducing preeclampsia models.
  • Exhibits a purity of 99.89%.