Learn More
Medchemexpress LLC L-NAME (hydrochloride) | 51298-62-5 | 99.9% | 269.69 | 5 G
SDP
Catalog No. 5000388243
Encompass_Preferred
L-NAME hydrochloride is an inhibitor of nitric oxide synthase (NOS) with an IC50 of 70 μM. It acts as a precursor to the NOS inhibitor L-NOARG, which has an IC50 value of 1.4 μM. It is used for research purposes only and is not sold to patients.
  • Inhibits NOS with an IC50 of 70 μM.
  • Precursor to the more potent NOS inhibitor L-NOARG (IC50 = 1.4 μM).
  • Can be used to induce hypertension models in animals.
  • Can be used to induce preeclampsia models in animals.
Catalog No. 50-003-88243 Supplier Medchemexpress LLC Supplier No. HY18729A5G
L-NAME hydrochloride is an inhibitor of nitric oxide synthase (NOS) with an IC50 of 70 μM. It acts as a precursor to the NOS inhibitor L-NOARG, which has an IC50 value of 1.4 μM. It is used for research purposes only and is not sold to patients.
  • Inhibits NOS with an IC50 of 70 μM.
  • Precursor to the more potent NOS inhibitor L-NOARG (IC50 = 1.4 μM).
  • Can be used to induce hypertension models in animals.
  • Can be used to induce preeclampsia models in animals.